missing translation for 'onlineSavingsMsg'
Learn More

Silver tetrafluoroborate, 99%

Product Code. p-4407832
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337237 10 g 10g
11327237 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337237 Supplier Thermo Scientific Alfa Aesar Supplier No. 011539.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Light Sensitive and Hygroscopic | CAS: 14104-20-2 | AgBF4 | 194.67 g/mol

Silver tetrafluoroborate finds applications as a useful reagent in inorganic, organic and organomettalic chemistry. It acts as a moderately strong oxidant in dichloromethane, replaces halide anions and ligands with weakly coordinating tetrafluoroborate anions, and behaves as a catalyst for cyclization reactions. It is a potent promoter for chemical glycosylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14104-20-2
Molecular Formula AgBF4
Molecular Weight (g/mol) 194.67
MDL Number MFCD00003408
InChI Key CCAVYRRHZLAMDJ-UHFFFAOYSA-N
Synonym silver tetrafluoroborate, silver borofluoride, silver fluoborate, silver fluoroborate, silver i tetrafluoroborate, borate 1-, tetrafluoro-, silver 1+, silver tetrafluroborate, acmc-209clm
PubChem CID 159722
IUPAC Name silver;tetrafluoroborate
SMILES [Ag+].F[B-](F)(F)F

Specifications

Melting Point 200°C (decomposition)
Color Brown to White
Physical Form Crystalline
Quantity 10 g
Assay Percent Range 99%
Solubility Information Soluble in water and polar organic solvents.
Formula Weight 194.67
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Appearance White to pale brown crystals or powder
Chemical Name or Material Silver tetrafluoroborate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.