missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver, Technical, Metal, Powder, SLR, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
10 g
Packaging:
Vial
Unit Size:
10g
Specifications
Specifications
| Color | Silver |
| CAS | 7440-22-4 |
| Melting Point | 960.5°C |
| Boiling Point | 2212°C |
| Quantity | 10 g |
| Packaging | Vial |
| Form | Solid |
| Molecular Formula | Ag |
| MDL Number | MFCD00003397 |
| Synonym | argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction