missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfide, 99.999% (metals basis)

Product Code. 15442277
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442277 25 g 25g
15432277 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15442277 Supplier Thermo Scientific Alfa Aesar Supplier No. 045458.14
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21548-73-2 | Ag2S | 247.80 g/mol

Silver sulfide is used as photosensitizer in photography, anti-bacterial and anti-microbial agents. It is also useful as qualitative inorganic analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21548-73-2
Molecular Formula Ag2S
Molecular Weight (g/mol) 247.80
MDL Number MFCD00003406
InChI Key XUARKZBEFFVFRG-UHFFFAOYSA-N
IUPAC Name disilver(1+) sulfanediide
SMILES [S--].[Ag+].[Ag+]

Specifications

Melting Point 825°C
Color Gray to Black
Physical Form Pieces
Quantity 25 g
UN Number UN3077
Merck Index 14,8530
Solubility Information Insoluble in water.
Formula Weight 247.8
Percent Purity 99.999%
Odor Odorless
Sensitivity Light sensitive
Density 7.317 g/mL
Assay (metals basis)
Chemical Name or Material Silver sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.