Learn More
Silver sulfide, Premion™, 99.995% (metals basis), Thermo Scientific Chemicals
Description
It finds use as a photosensitizer in photography. Blackening the silver for jewelleries and disinfection/sterilization of reservoirs. Anti-bacterial and anti-microbial agents. Laboratory reagents. A mixed ionic-electronic conductor. In IR detectors, photoconductors, photovoltaic cells, electrochemical storage cells, etc. Black colorant in ceramic glazes and for niello metal work. Autometallography in the detection of catalytic tissue metals and colloidal gold particles bound to macromolecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 825°C |
Color | Gray to Black |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.998% (metals basis) |
Solubility Information | Insoluble in most of the solvents. Insoluble in acids,alkalies and aqueous ammoniums. |
Formula Weight | 247.8 |
Odor | Odorless |
Sensitivity | Light sensitive |
Appearance | Dark grey to black powder/flake/crystalline/beads |
Show More |
RUO – Research Use Only