missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfide, 99.9+% (metals basis)

Product Code. 11413790
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413790 25 g 25g
11403790 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413790 Supplier Thermo Scientific Alfa Aesar Supplier No. 089473.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21548-73-2 | Ag2S | 247.80 g/mol

Silver sulfide is useful as a photosensitizer in photography, Intermediates, chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21548-73-2
Molecular Formula Ag2S
Molecular Weight (g/mol) 247.80
MDL Number MFCD00003406
InChI Key XUARKZBEFFVFRG-UHFFFAOYSA-N
IUPAC Name disilver(1+) sulfanediide
SMILES [S--].[Ag+].[Ag+]

Specifications

Melting Point 825°C
Color Gray to Black
Physical Form Crystalline Powder
Quantity 25 g
UN Number UN3077
Merck Index 14,8530
Solubility Information Soluble in aq. HCN,aq. citric acid with KNO3. Insoluble in acids,alkalies,aqueous ammoniums.
Formula Weight 247.8
Percent Purity ≥99.9%
Odor Odorless
Sensitivity Light sensitive
Density 7.317 g/mL
Assay (metals basis)
Chemical Name or Material Silver sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.