missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfate-Sulfuric acid solution, Volumetric solution, for COD determination according to DIN 38409, part 41, Honeywell Fluka™

Product Code. 15624320 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15624320

Brand: Honeywell-Fluka 346294X2.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Volumetric solution, for COD determination according to DIN 38409, part 41

  • Solutions are prepared with raw materials which meet or exceed requirements of the American Chemical Society
  • Where no ACS specifications exist, chemicals of highest purity available are used
  • All aqueous solutions are prepared using high purity, deionized, water which meets requirements for ASTM type 1 Reagent Grade Water
  • Exacting production and quality control records are maintained providing traceability of raw materials
  • All solutions are guaranteed to meet label specifications
  • All solution labels meet or exceed all ISO, DOT and OSHA regulations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9, 7732-18-5, 10294-26-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Boiling Point 338°C
CAS Min % ≥90.0000%, ≥1.0000%
CAS Max % ≤100.0000%, <5.0000%
Quantity 4 x 2.5 L
Linear Formula (CH3CH2)2O
UN Number UN3264
Packaging Glass Bottle
Density 1.85 g/cm3
Chemical Name or Material Silver sulfate-Sulfuric acid solution, Volumetric solution, for COD determination according to DIN 38409, part 41
compliance-icons compliance-icons
Product Identifier
  • Silver sulfate-Sulfuric acid solution
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, Disilver(1+) sulphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.