missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Sulfate, Certified AR for Analysis, Fisher Chemical™
£216.24 - £4218.95
Chemical Identifiers
CAS | 10294-26-5 |
---|---|
Molecular Formula | Ag2H2O4S |
Molecular Weight (g/mol) | 313.81 |
MDL Number | 3407 |
InChI Key | YPNVIBVEFVRZPJ-UHFFFAOYSA-N |
Synonym | silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt |
PubChem CID | 159865 |
IUPAC Name | disilver(1+) sulfuric acid |
SMILES | [Ag+].[Ag+].OS(O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10090340
|
Fisher Chemical
S/1400/46 |
25 g | Glass Jar | ||||||
10774381
|
Fisher Chemical
S/1400/48 |
100 g | Glass Jar |
£708.21
100g Qty Price Break Available |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11448822
|
Fisher Chemical
S/1400/53 |
500 g | Amber glass bottle |
£4218.95
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
10294-26-5 | |
313.81 | |
YPNVIBVEFVRZPJ-UHFFFAOYSA-N | |
159865 | |
[Ag+].[Ag+].OS(O)(=O)=O |
Ag2H2O4S | |
3407 | |
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt | |
disilver(1+) sulfuric acid |
Specifications
1085°C | |
10294-26-5 | |
5 to 6 | |
Ag2H2O4S | |
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt | |
[Ag+].[Ag+].OS(O)(=O)=O | |
313.81 | |
311.79g/mol |
652°C | |
White | |
Solid | |
3407 | |
YPNVIBVEFVRZPJ-UHFFFAOYSA-N | |
disilver(1+) sulfuric acid | |
159865 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Silver sulfate
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.