missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfate, ACS, 98% min, Thermo Scientific Chemicals

Catalog Number 11351239
Click to view available options
Quantity:
25 g
100 g
10294-26-5
Ag2O4S
311.792
MFCD00003407
YPNVIBVEFVRZPJ-UHFFFAOYSA-L
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
159865
disilver;sulfate
[O-]S(=O)(=O)[O-].[Ag+].[Ag+]
10294-26-5
Ag2O4S
311.792
MFCD00003407
YPNVIBVEFVRZPJ-UHFFFAOYSA-L
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
159865
disilver;sulfate
[O-]S(=O)(=O)[O-].[Ag+].[Ag+]

Silver(I) sulfate is employed in silver plating, and in electrolysis. It is a useful non-staining substitute to silver nitrate. Silver sulfate/iodine is an excellent iodination reagent for alkyl, alkoxybenzenes, aromatic amines and uridines. It is used in batteries; its pharmaceitucal composition is used as an antimicrobial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1085°C (decomposition)
Melting Point 657°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 98%
Solubility Information Partially soluble in water. Water solubility increases with temperature. Soluble in HNO3,ammonia and conc. H2SO4.
Formula Weight 311.8
Grade ACS Reagent
Sensitivity Light sensitive
Density 5.45 g/mL
Chemical Name or Material Silver sulfate, For AAS analysis
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'