missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfate, ACS reagent, Thermo Scientific Chemicals

Product Code. 13266309
Click to view available options
Quantity:
10 g
100 g
500 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-26-5
Molecular Formula Ag2H2O4S
Molecular Weight (g/mol) 313.81
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
Synonym silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
PubChem CID 159865
IUPAC Name disilver(1+) sulfuric acid
SMILES [Ag+].[Ag+].OS(O)(=O)=O

Specifications

Boiling Point 1085.0°C
Melting Point 652.0°C
Quantity 100 g
Linear Formula Ag2SO4
Fieser 01,1015; 03,254; 04,435
Merck Index 15,8668
Solubility Information Solubility in water: 7.4g/L (20°C). Other solubilities: soluble in nh4oh, nitric acid and sulfuric acid, insoluble in alcohol
Formula Weight 311.79
Grade ACS Reagent
Chemical Name or Material Silver sulfate
compliance-icons compliance-icons
Product Identifier
  • Silver sulfate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'