missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfadiazine, 98%, Thermo Scientific Chemicals

Product Code. 15579493 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15579493 25 g 25g
15569493 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15579493 Supplier Thermo Scientific Acros Supplier No. 459650250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22199-08-2
Molecular Formula C10H9AgN4O2S
Molecular Weight (g/mol) 357.14
InChI Key UEJSSZHHYBHCEL-UHFFFAOYSA-N
Synonym silver sulfadiazine, sulfadiazine silver, flamazine, silver sulphadiazine, silvadene, sulfadiazine silver salt, dermazin, thermazene, silver sulfadiazinate, sulfadiazine, silver
PubChem CID 441244
ChEBI CHEBI:9142
IUPAC Name silver;(4-aminophenyl)sulfonyl-pyrimidin-2-ylazanide
SMILES C1=CN=C(N=C1)[N-]S(=O)(=O)C2=CC=C(C=C2)N.[Ag+]

Specifications

Melting Point 285.0°C
Quantity 25 g
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 357.14
Percent Purity 98%
Infrared Spectrum Authentic
Loss on Drying 0.5% max.
Chemical Name or Material Silver sulfadiazine
compliance-icons
Product Identifier
  • Silver sulfadiazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.