missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Silver Protein for Histology, Strong

Product Code. 11375018
Change view
Click to view available options
Quantity:
5g
25g
110g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375018 5g 5g
11365018 25g 25g
11385018 110g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11375018 Supplier Thermo Scientific Chemicals Supplier No. 043970.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 9008-42-8

Silver protein is used in electron microscopy with periodic acid and thiocarbohydrazide or thiosemicarbohydrazide as a positive stain for carbohydrates such as glycogen. It can also be used for light microscopy to stain nerve tissue. It is also used in histological stain for microscopic analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9008-42-8
Synonym Protargol

Specifications

Color Pale yellow to brown
Form Powder
Format Powder
For Use With (Application) Histological stain for microscopic analysis
Product Type Silver Protein
Quantity 5g
Sensitivity Hygroscopic; Light sensitive

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.