missing translation for 'onlineSavingsMsg'
Learn More

Silver perchlorate hydrate, 99%

Product Code. 10403785 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10403785 100 g Glass bottle 100g
10440871 5 g Glass bottle 5g
10296450 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10403785 Supplier Thermo Scientific Acros Supplier No. 210211000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14242-05-8 | AgClO4 | 207.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14242-05-8
Molecular Formula AgClO4
Molecular Weight (g/mol) 207.31
MDL Number MFCD00149128
InChI Key YDHABVNRCBNRNZ-UHFFFAOYSA-M
Synonym silver perchlorate hydrate, silver perchlorate monohydrate, silver perchlorite, silver perchloric acid, acmc-1brvv, ag.clo4.h2o, silver i perchlorate monohydrate, silver 1+ ion hydrate perchlorate, silver 1+ hydrate perchlorate
PubChem CID 15959344
SMILES [Ag+].[O-][Cl](=O)(=O)=O

Specifications

Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Assay Percent Range 98.5% min. (on dry substance) (Argentometry)
Linear Formula AgClO4·xH2O
Merck Index 10, 8358
Solubility Information Solubility in water: 557g/100mL. Other solubilities: soluble in many organic solvents
IUPAC Name silver(1+) perchlorate
Formula Weight 207.32
Percent Purity 99%
Packaging Glass bottle
Chemical Name or Material Silver perchlorate hydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Silver perchlorate hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.