missing translation for 'onlineSavingsMsg'
Learn More

Silver perchlorate, anhydrous

Product Code. 11389857
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389857 5 g 5g
11369857 25 g 25g
11379857 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11389857 Supplier Thermo Scientific Alfa Aesar Supplier No. 042209.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Light Sensitive & Hygroscopic | CAS: 7783-93-9 | AgClO4 | 207.314 g/mol

Silver perchlorate is used as a catalyst in organic chemistry. It is an effective reagent for replacing halides ligands with perchlorate. It is also used in the manufacturing of explosives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-93-9
Molecular Formula AgClO4
Molecular Weight (g/mol) 207.314
MDL Number MFCD00003400
InChI Key YDHABVNRCBNRNZ-UHFFFAOYSA-M
Synonym silver perchlorate, unii-sv4x1u113o, silver 1+ ion perchlorate, perchloric acid, silver 1+ salt, silver 1+ salt, ag.clo4, perchloric acid, silver 1+ salt 1:1, silver 1+ perchlorate ion, silver i perchloratehydrate, silver i perchlorate, anhydrous
PubChem CID 24562
IUPAC Name silver;perchlorate
SMILES [O-]Cl(=O)(=O)=O.[Ag+]

Specifications

Melting Point 485°C (decomposition)
Color White
Physical Form Crystals
Quantity 5 g
Solubility Information Soluble in water and many organic solvents (benzene,toluene,pyridine)Soluble in water,benzene and toluene.
Formula Weight 207.32
Sensitivity Hygroscopic; Light sensitive
Appearance White crystals
Density 2.806 g/cm3 at 20°C
Chemical Name or Material Silver perchlorate
compliance-icons compliance-icons
Product Identifier
  • silver perchlorate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contain: Perchloric acid, silver(1+) salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.