missing translation for 'onlineSavingsMsg'
Learn More

Silver Oxide, Extra Pure, SLR, Fisher Chemical™

Product Code. 10315910 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
10315910 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 10315910

Brand: Fisher Chemical S/1320/46

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 20667-12-3
Molecular Formula Ag2O
Molecular Weight (g/mol) 231.74
MDL Number MFCD00003404
InChI Key KHJDQHIZCZTCAE-UHFFFAOYSA-N
IUPAC Name (argentiooxy)silver
SMILES [Ag]O[Ag]

Specifications

Color Black
Physical Form Powder/Solid
Quantity 25 g
Formula Weight 231.74g/mol
Packaging Glass Jar
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Silver(I) oxide
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.