missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver nitrite, 99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Silver nitrite is used in the silver plating process, hair dye, inks, permanent marker pens and determination of chloride ions in water and drilling fluids. It is used as Tollen’s reagent and used in the formation of aniline and nitro compounds in victor mayer reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 140°C (decomposition) |
Color | White |
Physical Form | Crystalline |
Quantity | 10 g |
Assay Percent Range | 99% (metals basis) |
Solubility Information | Soluble in hot water.Insoluble in ethanol. |
Formula Weight | 153.88 |
Odor | Odorless |
Sensitivity | Light sensitive |
Appearance | Light yellow crystals |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'