missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrite, 99%

Product Code. 10772305 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10772305

Brand: Thermo Scientific Acros 210690500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-99-5 | AgNO2 | 153.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-99-5
Molecular Formula AgNO2
Molecular Weight (g/mol) 153.88
MDL Number MFCD00003413
InChI Key KKKDGYXNGYJJRX-UHFFFAOYSA-M
Synonym silver nitrite, unii-t3mz57ogif, t3mz57ogif, silbernitrit, nitrous acid, silver 1+ salt, silver i nitrite, silvernitrite, acmc-20ajq7, ag.no2, silver nitrite 10g
PubChem CID 160904
IUPAC Name silver;nitrite
SMILES N(=O)[O-].[Ag+]

Specifications

Melting Point 140.0°C
Color Yellow to Green
Physical Form Powder and/or Agglomerates
Quantity 50 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula AgNO2
Fieser 01,1011; 06,515; 11,467
Merck Index 15, 8658
Solubility Information Solubility in water: 3.3g/L (20°C). Other solubilities: decomposes in diluted acids,insoluble in alcohol
Formula Weight 153.88
Percent Purity 99%
Packaging Glass bottle
Specific Gravity 4.45
Density 4.4500g/mL
Chemical Name or Material Silver nitrite
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.