missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate Solution, volumetric, 0.1M AgNO3 (0.1N), Honeywell Fluka™

Product Code. p-7120763 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
2 L
Packaging:
Plastic Bottle
Unit Size:
2 L
500mL
This item is not returnable. View return policy

Product Code. 15613570

Brand: Honeywell-Fluka 319430500ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Volumetric, 0.1 M AgNO3 (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver;nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Boiling Point 100°C
Melting Point <0°C
Quantity 500 mL
UN Number UN3264
Formula Weight 169.87g/mol
Packaging Plastic Bottle
Density 1.015g/mL at 25°C; Vapor Density: 5.8 (vs air)
Chemical Name or Material Silver Nitrate Solution, Volumetric, 0.1M AgNO3 (0.1N)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.