missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate Solution, volumetric, 0.1M AgNO3 (0.1N), Solstice

Product Code. 15613570 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
2 L
Packaging:
Plastic Bottle
Unit Size:
2 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15613570 500 mL Plastic Bottle 500mL
15603570 2 L Plastic Bottle 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15613570 Supplier Honeywell-Fluka Supplier No. 319430500ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Volumetric, 0.1 M AgNO3 (0.1N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver;nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Boiling Point 100°C
Melting Point <0°C
Quantity 500 mL
UN Number UN3264
Formula Weight 169.87g/mol
Packaging Plastic Bottle
Density 1.015g/mL at 25°C; Vapor Density: 5.8 (vs air)
Chemical Name or Material Silver Nitrate Solution, Volumetric, 0.1M AgNO3 (0.1N)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.