missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate Solution, 1.0M, Honeywell Fluka™
Volumetric, 1.0 M AgNO3 (1.0N)
£199.63 - £334.87
Chemical Identifiers
CAS | 7761-88-8 |
---|---|
Molecular Formula | AgNO3 |
Molecular Weight (g/mol) | 169.87 |
MDL Number | MFCD00003414 |
InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
PubChem CID | 24470 |
ChEBI | CHEBI:32130 |
IUPAC Name | silver(1+) nitrate |
SMILES | [Ag+].[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15633570
|
Honeywell-Fluka
319449-500ML |
500 mL |
£199.63
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15623570
|
Honeywell-Fluka
319449-1L |
1 L |
£334.87
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
7761-88-8 | |
169.87 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
24470 | |
silver(1+) nitrate |
AgNO3 | |
MFCD00003414 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
CHEBI:32130 | |
[Ag+].[O-][N+]([O-])=O |
Specifications
7761-88-8 | |
MFCD00003414 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
1.0M | |
Silver Nitrate Solution |
AgNO3 | |
3264 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
24470 | |
169.87g/mol | |
1.135g/mL at 25°C; Vapor Density: 5.8 (vs air) |
Safety and Handling
P220-P273-P280-P305 + P351 + P338-P310-P501
H314-H410