missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Usp, Fisher Chemical™

Product Code. p-7073175 Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
10 L
25 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
Unit Size:
10 L
2.5 L
25 L
This item is not returnable. View return policy

Product Code. 10111910

Brand: Fisher Chemical J/7330/17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

AgNO3, CAS Number-7761-88-8, silver nitrate, lunar caustic, argerol, silver mononitrate, silvernitrate, silver i nitrate, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, 2.5L, CHEBI:32130, Colorless, SQGYOTSLMSWVJD-UHFFFAOYSA-N, silver;nitrate, 3414, 169.872

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
Grade pH.Eur.
Packaging HDPE plastic bottle
Quantity 2.5 L
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Silver nitrate, 0.1M solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : Silver nitrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.