missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate Solution 0.0282M (0.0282N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mL
1 L
2.5 L
Packaging:
Amber glass bottle
HDPE plastic bottle
Specifications
Color | Colorless |
IUPAC Name | silver;nitrate |
Packaging | Amber glass bottle |
Quantity | 250 mL |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Silver nitrate 0.0282M
- Warning
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'