missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, ACS Reagent, ≥99.0%, Solstice

Product Code. 15601890 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Poly bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15601890 25 g Glass Bottle 25g
15691880 100 g Glass Bottle 100g
15611890 500 g Poly bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15601890 Supplier Honeywell-Fluka Supplier No. 20913925G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Melting Point 212°C (decomposition)(literature)
CAS Max % ≤100.0000%
Quantity 25 g
Linear Formula AgNO3
UN Number UN1493
IUPAC Name silver;nitrate
Formula Weight 169.87
Percent Purity >99%
Grade ACS Reagent
Packaging Glass Bottle
Density 4.350 g/cm3; Vapor Density: 5.8 (vs air)
Chemical Name or Material Silver nitrate, ACS Reagent, ≥99.0%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Silver nitrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Silver nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.