Learn More
Silver nitrate, ACS, 99.9+% (metals basis), Thermo Scientific Chemicals
Description
Silver nitrate is employed in Silver plating, photography, manufacturing of other silver compounds, mirrors, coloring porcelain, and staining in histology. It finds use as a stain in scanning electron microscopy, and a key ingredient in several silver-based antiseptic and antibacterial pharmaceutical compositions. Silver(I) nitrate is used to separate mixtures of alkenes by selective absorption, and for deprotection and oxidation reactions in organic synthesis. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 212°C |
Physical Form | Crystalline |
Quantity | 25 g |
Assay Percent Range | >99.9% (metals basis) |
Solubility Information | Freely soluble in water,alcohol and aqueous ammonia. Slightly soluble in ether. |
Formula Weight | 169.87 |
Grade | ACS Reagent |
Sensitivity | Light sensitive |
Density | 4.352 g/mL |
Chemical Name or Material | Silver nitrate, For AAS analysis |
Safety and Handling
- Silver nitrate
- Danger
- Oxidising solid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only