missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, Acculute Standard Volumetric Solution, Final Concentration 0.1N, Thermo Scientific Chemicals

Catalog Number 11339319
Click to view available options
Quantity:
1 Unit
6 Units
This item is not returnable. View return policy
7761-88-8
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
CHEBI:32130
silver(1+) nitrate
[Ag+].[O-][N+]([O-])=O
This item is not returnable. View return policy
7761-88-8
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
CHEBI:32130
silver(1+) nitrate
[Ag+].[O-][N+]([O-])=O

Silver nitrate 0.1N standard volumetric solution is used for volumetric analysis in laboratories. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 6 Units
Color Colorless
Formula Weight 169.87
Concentration or Composition (by Analyte or Components) Silver nitrate: 40%; Water: 60%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Solubility Miscible with water, alcohol and aqueous ammonia
Physical Form Liquid
Chemical Name or Material Silver nitrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'