missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, 99+%

Product Code. 11470990
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470990 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11470990 Supplier Thermo Scientific Alfa Aesar Supplier No. A13854.36
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 444°C
Melting Point 212°C
CAS 7761-88-8
Physical Form Crystalline
Quantity 500 g
Packaging Glass jar
Sensitivity Light Sensitive
Appearance White
Assay Percent Range 99+%
Decomposition Information >444°C
Density 4.352
Molecular Formula AgNO3
MDL Number MFCD00003414
UN Number UN1493
Merck Index 14,8518
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras
Solubility Information Freely soluble aq. soln., alcohol and aqueous ammonia. Slightly soluble in ether.
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES [Ag+].[O-][N+]([O-])=O
IUPAC Name silver(1+) nitrate
Molecular Weight (g/mol) 169.87
PubChem CID 24470
ChEBI CHEBI:32130
Formula Weight 169.87
Chemical Name or Material Silver nitrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.