missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silver needles, 99.999% (metals basis)

Brand:  Alfa Aesar™ 039697.09

Code : NEW

Additional Details : CAS Number : 7440-22-4 Weight : 0.01000kg Transport : UN number : 3077 Chem class : 9 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 11384657

  • £56.80 / 10g


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Chemical Identifiers

Silver needles
7440-22-4
MFCD00003397
UN3077

Specifications

Silver needles
99.999%
MFCD00003397
Characteristic
7440-22-4
(metals basis)
UN3077
10g
Videos
SDS