missing translation for 'onlineSavingsMsg'
Learn More

Silver methanesulfonate

Product Code. 11393447
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393447 50 g 50g
11383447 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393447 Supplier Thermo Scientific Alfa Aesar Supplier No. 018641.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2386-52-9 | CH3AgO3S | 202.96 g/mol

Silver methanesulfonate is used as a catalyst for heterocyclization reactions. Further, it is used in the synthesis of unsaturated ketones and esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2386-52-9
Molecular Formula CH3AgO3S
Molecular Weight (g/mol) 202.96
MDL Number MFCD00064795
InChI Key MLKQJVFHEUORBO-UHFFFAOYSA-M
Synonym silver methanesulfonate, silver methylsulfonate, silver methanesulphonate, methanesulfonic acid, silver salt, silver methanesulfonate, silver salt, silver i methanesulfonate, methanesulfonic acid, silver 1+ salt, silver methanesulfonate ag ch2so3, methylsulfonic acid silver salt, silvermesylate
PubChem CID 159658
IUPAC Name silver;methanesulfonate
SMILES CS(=O)(=O)[O-].[Ag+]

Specifications

Color White
Physical Form Powder
Quantity 50 g
Solubility Information Soluble in water,alcohol,ether and benzene. Slightly soluble in toluene,o-chlorotoluene and ethyl disulfide. Insoluble in hexane and methylcyclopentane.
Formula Weight 202.97
Odor Odorless
Sensitivity Light sensitive
Appearance White powder
Chemical Name or Material Silver methanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.