missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Silver Lake Research™ Well Water Test Kit

Product Code. 11755149 Shop All Cole-Parmer Products
Click to view available options
No. of Tests:
1/Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11755149

Brand: Cole-Parmer™ 9953004

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Silver Lake Research™ Well Water Test Kit provides lab-quality results in a simple-to-use, affordable kit.

  • Comprehensive test kits detect most common water contaminants.
  • Compare your results against EPA-recommended levels.
  • Fast, easy-to-use water test kits detect hazardous contaminants in municipal and private well drinking water.
  • Drinking water test kits indicate the presence of bacteria, pesticides and lead.
  • Kits also include readings for nitrate (0 to 50 ppm), nitrite (0 to 10 ppm), total chlorine (0 to 10 ppm), pH (6 to 10) and water hardness (0 to 425 ppm).
  • Well water test kits add tests for contaminants commonly found in private well water: iron and copper (0 to 5 ppm).
  • Order bacteria, pesticides, and lead stand-alone kits if less comprehensive tests are required.
  • All kits are for one-time use.

Specifications

pH Range 6 to 10
For Use With (Application) Detecting most common water contaminants.
No. of Tests 1/Kit
Test Method Reagent Strips
Type Drinking Water Test Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.