missing translation for 'onlineSavingsMsg'
Learn More

Silver lactate solution, Ag 4-5% w/w (cont. Ag), Thermo Scientific Chemicals

Product Code. 11366269
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366269 5 g 5g
11386269 1 g 1g
11376269 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11366269 Supplier Thermo Scientific Alfa Aesar Supplier No. 044858.NB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15768-18-0
Molecular Formula C3H5AgO3
Molecular Weight (g/mol) 196.938
MDL Number MFCD00043279
InChI Key LMEWRZSPCQHBOB-UHFFFAOYSA-M
Synonym silver lactate, silver lactate monohydrate, lactato-o1,o2 silver, lactic acid, silver salt, silver, 2-hydroxypropanoato-o1,o2, propanoic acid, 2-hydroxy-, silver complex, silver, 2-hydroxy-kappao propanoato-kappao
PubChem CID 91824762
IUPAC Name silver;2-hydroxypropanoate
SMILES CC(C(=O)[O-])O.[Ag+]

Specifications

Color Colorless
Linear Formula CH3CH(OH)CO2Ag
UN Number UN3082
Concentration Ag 4 to 5% w/w (cont. Ag)
Formula Weight 196.94
Quantity 5 g
Sensitivity Light sensitive
Physical Form Liquid
Chemical Name or Material Silver lactate solution

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.