missing translation for 'onlineSavingsMsg'
Learn More

Silver lactate, 96%

Product Code. 10226720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10226720 10 g Glass bottle 10g
10519833 5 g Glass bottle 5g
11974351 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10226720 Supplier Thermo Scientific Acros Supplier No. 233330100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15768-18- | C3H5O3Ag | 196.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15768-18-
Molecular Formula C3H5O3Ag
Molecular Weight (g/mol) 196.94
InChI Key LMEWRZSPCQHBOB-UHFFFAOYSA-M
Synonym silver lactate, silver lactate monohydrate, lactato-o1,o2 silver, lactic acid, silver salt, silver, 2-hydroxypropanoato-o1,o2, propanoic acid, 2-hydroxy-, silver complex, silver, 2-hydroxy-kappao propanoato-kappao
PubChem CID 91824762
IUPAC Name silver;2-hydroxypropanoate
SMILES CC(C(=O)[O-])O.[Ag+]

Specifications

Melting Point 120.0°C to 122.0°C
Color Brown-Gray to Gray
Physical Form Powder
Quantity 10 g
Assay Percent Range 95% min. (Argentometry)
Linear Formula CH3CH(OH)CO2Ag
Merck Index 14,8517
Solubility Information Solubility in water: 77g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 196.94
Percent Purity 96%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Silver lactate
Show More Show Less
compliance-icons
Product Identifier
  • Silver lactate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.