missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver iodide, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Used in medicine for its caustic, astringent and antiseptic effects. Used in photography, and in the estimation of silver. Through heterogeneous nucleation, it is used in cloud seeding, relying on its beta-AgI crystal structure being similar to that of Ice.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 558°C |
Physical Form | Crystalline |
Quantity | 100 g |
Assay Percent Range | 99.9% (metals basis) |
Solubility Information | Insoluble in water,acids and ammonium carbonate. Freely soluble in alkali cyanides and iodides. |
Formula Weight | 234.77 |
Sensitivity | Light sensitive |
Density | 5.67 g/mL |
Chemical Name or Material | Silver iodide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'