Learn More
Silver(II) oxide, 99.9% (metals basis), Ag 86.6% min
Description
Silver(II) oxide is used to make silver oxide-zinc alkali batteries. It is also used as a reagent in laboratory reactions and forms transition metal-carbene complexes. It is also used as a catalyst to prepare silver metal. It acts as an effective carbon dioxide scrubber and used in nuclear submarines, the international space station and space shuttles. Further, it is used in the manufacture of filters for gas sensors and in some infection-resistant surgical fabric materials and fibrous textile articles. In addition to this, it is used in concrete and in some swimming pools to protect the water from undesired microbes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >100°C (decomposition) |
| Color | Black to Gray |
| Physical Form | Powder |
| Quantity | 25 g |
| UN Number | UN3085 |
| Merck Index | 14,8522 |
| Solubility Information | Soluble in alkalis. Insoluble in water. |
| Formula Weight | 123.87 |
| Percent Purity | 99.9% |
| Concentration or Composition (by Analyte or Components) | Ag 86.6% min. |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.