missing translation for 'onlineSavingsMsg'
Learn More

Silver(II) oxide, 98%

Product Code. 11328087
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328087 100 g 100g
11338087 5 g 5g
11348087 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11328087 Supplier Thermo Scientific Alfa Aesar Supplier No. 022695.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufacturing of silver oxide-zinc alkali batteries | CAS: 1301-96-8 | AgO | 123.867 g/mol

Silver(II) oxide is used to make silver oxide-zinc alkali batteries. It is also used as a reagent in laboratory reactions and forms transition metal-carbene complexes. It is also used as a catalyst to prepare silver metal. It acts as an effective carbon dioxide scrubber and used in nuclear submarines, the international space station and space shuttles. Further, it is used in the manufacture of filters for gas sensors and in some infection-resistant surgical fabric materials and fibrous textile articles. In addition to this, it is used in concrete and in some swimming pools to protect the water from undesired microbes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1301-96-8
Molecular Formula AgO
Molecular Weight (g/mol) 123.867
MDL Number MFCD00044285
InChI Key OTCVAHKKMMUFAY-UHFFFAOYSA-N
Synonym silver ii oxide, silver oxide, silberperoxyd, silver oxide,ago, acmc-1aiyc, silver i,iii oxide, ksc203c6r, silver ii oxide 5g, silver ii oxide-ag
PubChem CID 92152
IUPAC Name oxosilver
SMILES O=[Ag]

Specifications

Melting Point >100°C (decomposition)
Color Black to Gray
Physical Form Powder
Quantity 100 g
UN Number UN3085
Merck Index 14,8522
Solubility Information Insoluble in water; Soluble in alkalies,ammonium hydroxide with decomposition and N2 formation.
Formula Weight 123.87
Percent Purity 98%
Odor Odorless
Sensitivity Light and moisture sensitive
Density 7.44 g/mL
Chemical Name or Material Silver(II) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.