missing translation for 'onlineSavingsMsg'
Learn More

Silver(II) fluoride, 98+%

Product Code. 11397357
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397357 50 g 50g
11377357 2 g 2g
11387357 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397357 Supplier Thermo Scientific Alfa Aesar Supplier No. 011610.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Moisture and Light Sensitive | CAS: 7783-95-1 | AgF2

Silver(II) fluoride is used as a fluorinating and oxidizing agent. It is also used in the preparation of organic perfluorocompounds and in the fluorination of aromatic compounds. In addition, it is also used in pharmacy and electroplating industry. It serves as an intermediate in the catalysis of gaseous reactions with fluorine by silver. It is utilized in the oxidation of xenon to xenon difluoride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-95-1
Molecular Formula AgF2
MDL Number MFCD00003411
Synonym Silver difluoride

Specifications

Melting Point 690°C
Color Brown
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 98+%
Solubility Information Soluble in hexane,chloroform,chlorofluorocarbon and hydrogen fluoride.
Formula Weight 145.87
Sensitivity Light and moisture sensitive
Appearance Brown crystals
Density 4.6
Chemical Name or Material Silver(II) fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.