missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) Oxide, TRC

Product Code. 30240507
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240507 25 g 25g
30240504 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30240507 Supplier Toronto Research Chemicals Supplier No. TRCS46505325G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20667-12-3
Molecular Formula Ag2O
Molecular Weight (g/mol) 231.74
Synonym Argentous Oxide, CF 01, Disilver Monoxide, Disilver Oxide, Silver Oxide, Silver(I) Oxide
IUPAC Name argentiooxysilver
SMILES [Ag]O[Ag]

Specifications

Quantity 25 g
InChI Formula InChI=1S/2Ag.O
Formula Weight 229.8051
Chemical Name or Material Silver(I) Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.