missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) Oxide, purum p.a, ≥99.0% (AT), Solstice

Product Code. 15627350 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Poly Bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15627350 50 g Poly Bottle 50g
15617350 10 g Poly Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15627350 Supplier Honeywell Research Chemicals Supplier No. 8526050G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥99.0% (AT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20667-12-3
Molecular Formula Ag2O
Molecular Weight (g/mol) 231.74
MDL Number MFCD00003404
InChI Key KHJDQHIZCZTCAE-UHFFFAOYSA-N
IUPAC Name (argentiooxy)silver
SMILES [Ag]O[Ag]

Specifications

Color Gray
Physical Form Powder
Quantity 50 g
UN Number UN1479
Solubility Information Slightly soluble
Formula Weight 231.74g/mol
Percent Purity ≥99.0% (AT)
Odor Odorless
Packaging Poly Bottle
Chemical Name or Material Silver(I) oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.