missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) oxide, 99+% (metals basis)

Product Code. 11371209
Click to view available options
Quantity:
25 g
500 g
Unit Size:
25g
500g
This item is not returnable. View return policy

Product Code. 11371209

Brand: Thermo Scientific Alfa Aesar 011407.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20667-12-3 | Ag2O | 231.74 g/mol

Polishing glass, coloring glass yellow, catalyst, purifying drinking water, lab reagent, carbon dioxide scrubber, and chemical sensors. It is used in the preparation of other silver compounds, and silver-oxide batteries. In organic chemistry, silver oxide finds use as an oxidizing agent for aldehyes to produce carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20667-12-3
Molecular Formula Ag2O
Molecular Weight (g/mol) 231.74
MDL Number MFCD00003404
InChI Key KHJDQHIZCZTCAE-UHFFFAOYSA-N
IUPAC Name (argentiooxy)silver
SMILES [Ag]O[Ag]

Specifications

Melting Point 230°C (decomposition)
Physical Form Powder
Quantity 500 g
Assay Percent Range >99% (metals basis)
Solubility Information Soluble in dilute HNO3 and ammonia. Practically insoluble in water. Insoluble in alcohol
Formula Weight 231.74
Sensitivity Light sensitive
Density 7.2 g/mL
Chemical Name or Material Silver(I) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.