missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) fluoride, Puriss. p.a., ≥99.0%, Honeywell™ Fluka™

Product Code. 15677340 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Poly Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15677340 5 g Poly Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15677340 Supplier Honeywell-Fluka Supplier No. 851845G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a.,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 1150°C
Melting Point 435°C
CAS 7775-41-9
Quantity 5 g
Molecular Formula AgF
MDL Number MFCD00003410
UN Number UN3260
Synonym silver fluoride, silver 1+ fluoride, silver 1+ ion fluoride, argentous fluoride, ag-f, silverfluoride, silver salt, fluoride, silver fluoride,, silver i fluoride 5g
InChI Key REYHXKZHIMGNSE-UHFFFAOYSA-M
SMILES [F-].[Ag+]
IUPAC Name silver;fluoride
Molecular Weight (g/mol) 126.867
PubChem CID 165912
ChEBI CHEBI:30340
Formula Weight 126.87g/mol
Percent Purity ≥99.0%
Packaging Poly Bottle
Density 5.852g/mL at 25°C
Chemical Name or Material Silver(I) fluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.