missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) fluoride, 99+%

Product Code. 10531081 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Plastic bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10531081 25 g Plastic bottle 25g
10022630 5 g Plastic bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10531081 Supplier Thermo Scientific Acros Supplier No. 210450250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7775-41-9 | AgF | 126.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-41-9
Molecular Formula AgF
Molecular Weight (g/mol) 126.87
MDL Number MFCD00003410
InChI Key REYHXKZHIMGNSE-UHFFFAOYSA-M
Synonym silver fluoride, silver 1+ fluoride, silver 1+ ion fluoride, argentous fluoride, ag-f, silverfluoride, silver salt, fluoride, silver fluoride,, silver i fluoride 5g
PubChem CID 165912
ChEBI CHEBI:30340
IUPAC Name silver;fluoride
SMILES [F-].[Ag+]

Specifications

Boiling Point 1150.0°C
Melting Point 435.0°C
Color Brown-Rust to Brown-Yellow
Physical Form Powder
Quantity 25 g
Assay Percent Range 99% min
Linear Formula AgF
Fieser 02,364; 05,581; 06,514; 15,287
Merck Index 15, 8653
Solubility Information Solubility in water: in water: 182 g/100mL (15.5°C).
Formula Weight 126.87
Percent Purity ≥99%
Packaging Plastic bottle
Specific Gravity 5.85
Density 5.8500g/mL
Chemical Name or Material Silver(I) fluoride, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Silver(I) fluoride
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.