missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) fluoride, 98%

Product Code. 11367357
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367357 25 g 25g
11357357 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367357 Supplier Thermo Scientific Alfa Aesar Supplier No. 011609.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Light and Moisture Sensitive and Hygroscopic | CAS: 7775-41-9 | AgF | 126.867 g/mol

Silver(I) fluoride is used as a fluorination and desilylation reagent in organic synthesis. In medical filed, it is used as topical caries in aqueous form in dentistry. It is employed in organofluorine chemistry for addition of fluoride across multiple bonds, especially to prepare perfluoroalkylsilver(I) by reacting with alkenes. Further, it is involved in the preparation of tetralkylammonium fluorides by the reaction with tetralkylammonium bromide. In addition to this, it is used as a desulfuration-fluorination reagent on thiourea derived substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-41-9
Molecular Formula AgF
Molecular Weight (g/mol) 126.867
MDL Number MFCD00003410
InChI Key REYHXKZHIMGNSE-UHFFFAOYSA-M
Synonym silver fluoride, silver 1+ fluoride, silver 1+ ion fluoride, argentous fluoride, ag-f, silverfluoride, silver salt, fluoride, silver fluoride,, silver i fluoride 5g
PubChem CID 165912
ChEBI CHEBI:30340
IUPAC Name silver;fluoride
SMILES [F-].[Ag+]

Specifications

Boiling Point ca 1150°C
Melting Point 435°C
Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 98%
Solubility Information Soluble in HF,ammonia,CH3CNSoluble in water,hydrogen fluoride and methanol.
Formula Weight 126.87
Sensitivity Hygroscopic; Light and moisture sensitive
Appearance Orange-brown crystals
Density 5.852
Chemical Name or Material Silver(I) fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.