missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) citrate hydrate

Product Code. 15432317
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15432317

Brand: Thermo Scientific Alfa Aesar 045496.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 206986-90-5 | C6H5Ag3O7 | 512.70 g/mol

Silver(I) citrate hydrate can be very attractive for the future biomedical or pharmaceutical/therapeutic applications. It can be used as an antimicrobial agent and enhancers of antioxidants and anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206986-90-5
Molecular Formula C6H5Ag3O7
Molecular Weight (g/mol) 512.70
MDL Number MFCD00150589
InChI Key QUTYHQJYVDNJJA-UHFFFAOYSA-K
Synonym silver i citrate hydrate, trisilver 1+ citrate hydrate
PubChem CID 16212313
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;silver;hydrate
SMILES [Ag+].[Ag+].[Ag+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Melting Point 170°C (decomposition)
Quantity 100 g
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 512.70 (Anhydrous)
Physical Form Powder
Chemical Name or Material Silver(I) citrate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.