missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) behenate

Product Code. 15452307
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452307 5 g 5g
15462307 25 g 25g
15472307 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15452307 Supplier Thermo Scientific Alfa Aesar Supplier No. 045494.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2489-05-6 | C22H43AgO2 | 447.45 g/mol

Silver(I) behenate is used as a calibration standard in X-ray spectrophotometer to an absolute X-ray energy scale. It is utilized for low-angle diffraction standard in X-ray crystallography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2489-05-6
MDL Number MFCD00059001

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 5 g
Molecular Formula C22H43AgO2
Synonym silver behenate, silver docosanoate, behenic acid silver salt, unii-n3hp26f3pb, n3hp26f3pb, silver hebenate, docosanoic acid, silver 1+ salt 1:1, docosanoic acid silver salt, docosanoic acid, silver salt, docosanoic acid, silver 1+ salt
Solubility Information Slightly soluble in water.
InChI Key AQRYNYUOKMNDDV-UHFFFAOYSA-M
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)[O-].[Ag+]
IUPAC Name silver;docosanoate
Molecular Weight (g/mol) 447.45
PubChem CID 164971
Formula Weight 447.44
Sensitivity Light sensitive
Chemical Name or Material Silver(I) behenate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.