missing translation for 'onlineSavingsMsg'
Learn More

Silver hexafluorophosphate, 98%, -6 mesh

Código de producto. 10413265 Tienda Thermo Scientific Chemicals Productos
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Plastic bottle
Tamaño de la unidad:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Packaging unitSize
10413265 25 g Plastic bottle 25g
10206510 1 g Plastic bottle 1g
10104650 10 g Plastic bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10413265 Proveedor Thermo Scientific Acros N.º de proveedor 211120250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 26042-63-7 | AgF6P | 252.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 26042-63-7
Molecular Formula AgF6P
Molecular Weight (g/mol) 252.83
MDL Number MFCD00003415
InChI Key SCQBROMTFBBDHF-UHFFFAOYSA-N
Synonym silver hexafluorophosphate, silver hexafluorophosphate v, ag.f6p, silver i hexafluorophosphate, agpf6, acmc-1ceef, phosphate 1-, hexafluoro-, silver 1+, phosphate 1-, hexafluoro-, silver 1+ 1:1, silver i hexafluorophosphate v
PubChem CID 168464
IUPAC Name silver;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.[Ag+]

Especificaciones

Color Beige-Gray to White
Physical Form Powder
Quantity 25 g
Linear Formula AgPF6
Solubility Information Solubility in water: soluble
Formula Weight 252.83
Percent Purity 98%
Packaging Plastic bottle
Chemical Name or Material Silver hexafluorophosphate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.