missing translation for 'onlineSavingsMsg'
Learn More

Silver hexafluorophosphate, 98%

Product Code. 11391948
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11391948

Brand: Thermo Scientific Alfa Aesar 011873.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26042-63-7 | AgF6P | 252.832 g/mol

It is commonly used as an oxidant forming silver metal. It is often used as a reagent in inorganic and organometallic chemistry. It is used in the preparation of engineering novel materials as well as in preparation of supra molecular Ag(I) complexes. It is used in the development of new catalysts, replacing halide ligands with the weakly coordinating hexafluorophosphate anion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26042-63-7
Molecular Formula AgF6P
Molecular Weight (g/mol) 252.832
MDL Number MFCD00003415
InChI Key SCQBROMTFBBDHF-UHFFFAOYSA-N
Synonym silver hexafluorophosphate, silver hexafluorophosphate v, ag.f6p, silver i hexafluorophosphate, agpf6, acmc-1ceef, phosphate 1-, hexafluoro-, silver 1+, phosphate 1-, hexafluoro-, silver 1+ 1:1, silver i hexafluorophosphate v
PubChem CID 168464
IUPAC Name silver;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.[Ag+]

Specifications

Melting Point 102°C (decomposition)
Color Brown to Yellow
Physical Form Powder
Quantity 5 g
Assay Percent Range 98%
Solubility Information Soluble in organic solvents such as benzene,toluene,and acetonitrile.
Formula Weight 252.83
Odor Odorless
Sensitivity Light sensitive
Appearance White powder
Chemical Name or Material Silver hexafluorophosphate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.