missing translation for 'onlineSavingsMsg'
Learn More

Silver hexafluoroantimonate, 99%

Product Code. 11397227
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397227 25 g 25g
11307237 1 g 1g
11317237 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397227 Supplier Thermo Scientific Alfa Aesar Supplier No. 011538.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26042-64-8 | AgF6Sb | 343.619 g/mol

Silver hexafluoroantimonate is used as a catalyst for the preparation of a number of compounds. It acts as an acidic catalyst in epoxide opening reactions and sulfenylation reactions , controlled initiator in polymerization reactions, and as an activator of catalyst in certain organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26042-64-8
Molecular Formula AgF6Sb
Molecular Weight (g/mol) 343.619
MDL Number MFCD00003401
InChI Key GSXFODUDOAXUBF-UHFFFAOYSA-H
Synonym silver hexafluoroantimonate, silver hexafluoroantimonate v, silver hexafluorostiboranuide, silver 1+ hexafluorostibanuide, ag.f6sb, silverhexafluoroantimonate, agsbf6, acmc-209tq8, ksc492g5j, silver i hexafluoroantimonate
PubChem CID 16687962
IUPAC Name silver;hexafluoroantimony(1-)
SMILES F[Sb-](F)(F)(F)(F)F.[Ag+]

Specifications

Color White to Cream
Physical Form Lump/Powder
Quantity 25 g
UN Number UN3260
Solubility Information Soluble in water.
Formula Weight 343.41
Percent Purity 99%
Sensitivity Hygroscopic; Light sensitive
Chemical Name or Material Silver hexafluoroantimonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.