missing translation for 'onlineSavingsMsg'
Learn More

Silver cyanide, 99%

Product Code. 11371249
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371249 25 g 25g
11361249 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371249 Supplier Thermo Scientific Alfa Aesar Supplier No. 011424.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Silver plating | CAS: 506-64-9 | CAgN | 133.886 g/mol

Silver cyanide is used in silver-plating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-64-9
Molecular Formula CAgN
Molecular Weight (g/mol) 133.886
MDL Number MFCD00003409
InChI Key LFAGQMCIGQNPJG-UHFFFAOYSA-N
Synonym silver cyanide, silver cyanide ag cn, silver 1+ cyanide, kyanid stribrny czech, cyanure d'argent french, rcra waste number p104, silver i cyanide, rcra waste no. p104, kyanid stribrny, cyanure d'argent
PubChem CID 10475
IUPAC Name silver;cyanide
SMILES [C-]#N.[Ag+]

Specifications

Melting Point 320°C (decomposition)
Color White
Physical Form Powder
Quantity 25 g
Linear Formula AgCN
UN Number UN1684
Merck Index 14,8512
Solubility Information Insoluble in water,dilute acids,and alcohol; Soluble in alkali cyanides and boiling concentrated HNO3
Formula Weight 133.84
Percent Purity 99%
Odor Odorless
Refractive Index 1.685
Sensitivity Light sensitive
Density 3.95 g/mL
Chemical Name or Material Silver cyanide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.