missing translation for 'onlineSavingsMsg'
Learn More

Silver cyanide, 99%, for analysis

Product Code. 10752991 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10752991 10 g Glass bottle 10g
10042260 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10752991 Supplier Thermo Scientific Acros Supplier No. 180230100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 506-64-9 | CAgN | 133.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-64-9
Molecular Formula CAgN
Molecular Weight (g/mol) 133.89
MDL Number MFCD00003409
InChI Key LFAGQMCIGQNPJG-UHFFFAOYSA-N
Synonym silver cyanide, silver cyanide ag cn, silver 1+ cyanide, kyanid stribrny czech, cyanure d'argent french, rcra waste number p104, silver i cyanide, rcra waste no. p104, kyanid stribrny, cyanure d'argent
PubChem CID 10475
IUPAC Name silver;cyanide
SMILES [C-]#N.[Ag+]

Specifications

Melting Point >300°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 98.5% min
Linear Formula AgCN
Fieser 01,1006; 05,581; 07,320
Merck Index 15, 8651
Solubility Information Solubility in water: insoluble. Other solubilities: sol. in nh4oh,alkali cyanides,boiling conc. hno3,insoluble in alcohol and diluted acids
Formula Weight 133.89
Percent Purity 99%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Silver cyanide, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Silver cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.