missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >300°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 50 g |
| Assay Percent Range | 98.5% min |
| Linear Formula | AgCN |
| Fieser | 01,1006; 05,581; 07,320 |
| Merck Index | 15, 8651 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: sol. in nh4oh,alkali cyanides,boiling conc. hno3,insoluble in alcohol and diluted acids |
| Formula Weight | 133.89 |
| Show More |
Safety and Handling
Product Identifier
- Silver cyanide
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- H310-Fatal in contact with skin.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H330-Fatal if inhaled.
- H300-Fatal if swallowed.
- EUH032-Contact with acids liberates very toxic gas.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P361-Take off immediately all contaminated clothing.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?