missing translation for 'onlineSavingsMsg'
Learn More

Silver cyanate, 98%

Product Code. 15472257
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15472257 25 g 25g
15462257 5 g 5g
15482257 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15472257 Supplier Thermo Scientific Alfa Aesar Supplier No. 045411.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3315-16-0 | CHAgNO | 150.893 g/mol

Silver cyanate is a chemosterilant in adult house flies. It is also used as a reagent to prepare 1-(3-thianyl)uracil and 9-(3-thianyl)adenine derivatives, which has potential anti-human immunodeficiency virus (HIV) and antitumour activity.It is used as a silver plating solution in the electroplating process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3315-16-0
Molecular Formula CHAgNO
Molecular Weight (g/mol) 150.893
MDL Number MFCD00011512
InChI Key VAQVDZZAAMAZAO-UHFFFAOYSA-N
Synonym silver,cyanate, cyanic acid; silver
PubChem CID 45051745
IUPAC Name cyanic acid;silver
SMILES C(#N)O.[Ag]

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 98%
Beilstein 3655040
Solubility Information Soluble in ammonia,nitric acid,potassium cyanide,ammonium hydroxide. Insoluble in alcohol and dilute acids.
Formula Weight 149.89
Odor Odorless
Sensitivity Light sensitive
Density 4
Chemical Name or Material Silver cyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.