missing translation for 'onlineSavingsMsg'
Learn More

Silver cyanate, 98%, Thermo Scientific Chemicals

Product Code. 15462257
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Silver cyanate is a chemosterilant in adult house flies. It is also used as a reagent to prepare 1-(3-thianyl)uracil and 9-(3-thianyl)adenine derivatives, which has potential anti-human immunodeficiency virus (HIV) and antitumour activity.It is used as a silver plating solution in the electroplating process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3315-16-0
Molecular Formula CHAgNO
Molecular Weight (g/mol) 150.893
MDL Number MFCD00011512
InChI Key VAQVDZZAAMAZAO-UHFFFAOYSA-N
Synonym silver,cyanate, cyanic acid; silver
PubChem CID 45051745
IUPAC Name cyanic acid;silver
SMILES C(#N)O.[Ag]

Specifications

Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 98%
Beilstein 3655040
Solubility Information Soluble in ammonia,nitric acid,potassium cyanide,ammonium hydroxide. Insoluble in alcohol and dilute acids.
Formula Weight 149.89
Odor Odorless
Sensitivity Light sensitive
Density 4
Chemical Name or Material Silver cyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'